Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHLHE40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245010.1 | NP_001231939.1 | 4 - 5 | 652 | 61 | |
XM_021068423.1 | XP_020924082.1 | 758 | 61 | |||
XM_021068424.1 | XP_020924083.1 | 817 | 61 | |||
GenBank mRNA | AK345015.1 | - | 4 - 5 | 652 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons |