Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246243.1 | NP_001233172.1 | 2 - 3 | 695 | 97 | |
GenBank mRNA | GU929943.1 | - | 2 - 3 | 695 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |