Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PQBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246668.1 | NP_001233597.1 | 3 - 4 | 313 | 73 | |
XM_005673594.3 | XP_005673651.1 | 457 | 73 | |||
XM_013986114.1 | XP_013841568.1 | 723 | 73 | |||
GenBank mRNA | AK231983.1 | - | 4 - 5 | 449 | 73 | |
AK238508.1 | - | 4 - 5 | 728 | 73 | ||
AK352148.1 | - | 4 - 5 | 449 | 73 | ||
AK397584.1 | - | 4 - 5 | 452 | 73 | ||
AY609811.1 | - | 357 | 73 | |||
JF750401.1 | - | 3 - 4 | 313 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |