Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206401.1 | NP_001193330.1 | 4 - 5 | 572 | 94 | |
GenBank mRNA | CV874035.1 | - | 4 - 5 | 546 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |