Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206403.1 | NP_001193332.1 | 1 - 2 | 652 | 60 | |
GenBank mRNA | AK395814.1 | - | 1 - 2 | 652 | 60 | |
FS691891.1 | - | 1 - 2 | 652 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |