Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TADA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206536.1 | NP_001193465.1 | 4 - 5 | 745 | 85 | |
XM_021068275.1 | XP_020923934.1 | 720 | 85 | |||
GenBank mRNA | AK233113.1 | - | 4 - 5 | 704 | 85 | |
AK235953.1 | - | 4 - 5 | 745 | 85 | ||
AK238770.1 | - | 4 - 5 | 650 | 85 | ||
AK392946.1 | - | 4 - 5 | 873 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |