Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243056.1 | NP_001229985.1 | 12 - 13 | 1805 | 86 | |
XM_021093328.1 | XP_020948987.1 | 1912 | 86 | |||
XM_021093330.1 | XP_020948989.1 | 2011 | 86 | |||
XM_021093331.1 | XP_020948990.1 | 1757 | 86 | |||
XM_021093332.1 | XP_020948991.1 | 1733 | 86 | |||
XM_021093333.1 | XP_020948992.1 | 2207 | 86 | |||
XM_021093334.1 | XP_020948993.1 | 1929 | 86 | |||
XM_021093335.1 | XP_020948994.1 | 1787 | 86 | |||
XM_021093336.1 | XP_020948995.1 | 3343 | 86 | |||
XM_021093337.1 | XP_020948996.1 | 1966 | 86 | |||
XM_021093338.1 | XP_020948997.1 | 1881 | 86 | |||
GenBank mRNA | AK348726.1 | - | 12 - 13 | 1805 | 86 | |
AK349595.1 | - | 12 - 13 | 2024 | 86 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Sus scrofa Probe spans exons |