Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243185.1 | NP_001230114.1 | 2 - 3 | 257 | 71 | |
XM_021079065.1 | XP_020934724.1 | 841 | 71 | |||
GenBank mRNA | AK235554.1 | - | 2 - 3 | 235 | 71 | |
AK391745.1 | - | 2 - 3 | 237 | 71 | ||
BW981712.1 | - | 2 - 3 | 257 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |