Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243190.1 | NP_001230119.1 | 3 - 4 | 881 | 87 | |
XM_021085689.1 | XP_020941348.1 | 875 | 87 | |||
GenBank mRNA | AK345304.1 | - | 3 - 4 | 881 | 87 | |
AK394158.1 | - | 3 - 4 | 877 | 87 | ||
HF677512.1 | - | 3 - 4 | 623 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |