Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243190.1 | NP_001230119.1 | 4 - 5 | 1016 | 82 | |
XM_021085689.1 | XP_020941348.1 | 1010 | 82 | |||
GenBank mRNA | AK345304.1 | - | 4 - 5 | 1016 | 82 | |
HF677512.1 | - | 4 - 5 | 758 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |