Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHFPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243213.1 | NP_001230142.1 | 3 - 4 | 229 | 81 | |
XM_021079489.1 | XP_020935148.1 | 254 | 81 | |||
XM_021079500.1 | XP_020935159.1 | 1459 | 81 | |||
XM_021079502.1 | XP_020935161.1 | 527 | 81 | |||
XM_021079510.1 | XP_020935169.1 | 590 | 81 | |||
XM_021079514.1 | XP_020935173.1 | 283 | 81 | |||
XM_021079518.1 | XP_020935177.1 | 252 | 81 | |||
XM_021079527.1 | XP_020935186.1 | 419 | 81 | |||
GenBank mRNA | AK347793.1 | - | 3 - 4 | 206 | 81 | |
AK348362.1 | - | 3 - 4 | 229 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |