Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243289.1 | NP_001230218.1 | 1 - 2 | 508 | 90 | |
XM_021079380.1 | XP_020935039.1 | 563 | 90 | |||
GenBank mRNA | AK343621.1 | - | 1 - 2 | 508 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |