Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243291.1 | NP_001230220.1 | 1 - 2 | 180 | 77 | |
GenBank mRNA | AK237448.1 | - | 1 - 2 | 168 | 77 | |
AK390485.1 | - | 1 - 2 | 180 | 77 | ||
FS646397.1 | - | 1 - 2 | 180 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |