Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243341.1 | NP_001230270.1 | 2 - 3 | 290 | 125 | |
XM_005670048.3 | XP_005670105.1 | 1355 | 125 | |||
XM_021068495.1 | XP_020924154.1 | 1585 | 125 | |||
GenBank mRNA | AK235943.1 | - | 2 - 3 | 309 | 125 | |
BP463051.1 | - | 2 - 3 | 292 | 125 | ||
BW980083.1 | - | 2 - 3 | 251 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |