Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243352.1 | NP_001230281.1 | 2 - 3 | 558 | 128 | |
XM_005654033.3 | XP_005654090.1 | 3248 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |