Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243353.1 | NP_001230282.1 | 3 - 4 | 287 | 78 | |
GenBank mRNA | AK344967.1 | - | 3 - 4 | 287 | 78 | |
AK351608.1 | - | 3 - 4 | 286 | 78 | ||
AK400238.1 | - | 3 - 4 | 273 | 78 | ||
AK400822.1 | - | 3 - 4 | 287 | 78 | ||
FS641076.1 | - | 3 - 4 | 287 | 78 | ||
FS700194.1 | - | 3 - 4 | 284 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |