Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243367.1 | NP_001230296.1 | 4 - 5 | 936 | 112 | |
XM_021085767.1 | XP_020941426.1 | 728 | 112 | |||
GenBank mRNA | AK350557.1 | - | 4 - 5 | 936 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |