Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243382.1 | NP_001230311.1 | 1 - 2 | 60 | 87 | |
XM_005662132.3 | XP_005662189.1 | 599 | 87 | |||
GenBank mRNA | AK394033.1 | - | 1 - 2 | 43 | 87 | |
BW983759.1 | - | 1 - 2 | 60 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |