Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243387.1 | NP_001230316.1 | 2 - 3 | 476 | 97 | |
GenBank mRNA | AK231345.1 | - | 2 - 3 | 476 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |