Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243394.1 | NP_001230323.1 | 5 - 6 | 784 | 100 | |
GenBank mRNA | AK236877.1 | - | 5 - 6 | 784 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |