Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DESI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243397.1 | NP_001230326.1 | 4 - 5 | 478 | 105 | |
XM_005652508.3 | XP_005652565.1 | 547 | 105 | |||
GenBank mRNA | AK344907.1 | - | 4 - 5 | 481 | 105 | |
BP441548.1 | - | 4 - 5 | 478 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |