Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243405.1 | NP_001230334.1 | 1 - 2 | 194 | 104 | |
GenBank mRNA | AK235001.1 | - | 1 - 2 | 214 | 104 | |
AK346199.1 | - | 1 - 2 | 195 | 104 | ||
BP164995.1 | - | 1 - 2 | 194 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |