Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243410.1 | NP_001230339.1 | 5 - 6 | 797 | 85 | |
GenBank mRNA | AK344685.1 | - | 5 - 6 | 768 | 85 | |
AK389783.1 | - | 5 - 6 | 755 | 85 | ||
AK392309.1 | - | 5 - 6 | 798 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |