Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5H12orf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243412.1 | NP_001230341.1 | 13 - 14 | 1717 | 66 | |
NM_001243413.1 | NP_001230342.1 | 12 - 13 | 1574 | 66 | ||
XM_005655700.3 | XP_005655757.1 | 2357 | 66 | |||
XM_013988430.2 | XP_013843884.1 | 2442 | 66 | |||
XM_013988431.2 | XP_013843885.1 | 1616 | 66 | |||
XM_021090919.1 | XP_020946578.1 | 2363 | 66 | |||
XM_021090921.1 | XP_020946580.1 | 1606 | 66 | |||
XM_021090923.1 | XP_020946582.1 | 1431 | 66 | |||
XM_021090924.1 | XP_020946583.1 | 2168 | 66 | |||
GenBank mRNA | AK343836.1 | - | 13 - 14 | 1701 | 66 | |
AK400126.1 | - | 13 - 14 | 1700 | 66 | ||
EW203828.2 | - | 2 - 3 | 165 | 66 | ||
EW379696.2 | - | 5 - 6 | 498 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |