Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243414.1 | NP_001230343.1 | 2 - 3 | 174 | 116 | |
GenBank mRNA | AK239489.1 | - | 2 - 3 | 130 | 116 | |
BP464075.1 | - | 2 - 3 | 134 | 116 | ||
FD600058.1 | - | 2 - 3 | 174 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |