Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243418.1 | NP_001230347.1 | 9 - 10 | 1231 | 83 | |
XM_005655767.3 | XP_005655824.1 | 1236 | 83 | |||
XM_021093432.1 | XP_020949091.1 | 1338 | 83 | |||
GenBank mRNA | AK237952.1 | - | 9 - 10 | 1232 | 83 | |
AK239378.1 | - | 9 - 10 | 1203 | 83 | ||
AK392132.1 | - | 9 - 10 | 1227 | 83 | ||
AK398017.1 | - | 9 - 10 | 1224 | 83 | ||
AW785058.1 | - | 1 - 2 | 58 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |