Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCHFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243443.1 | NP_001230372.1 | 1 - 2 | 141 | 72 | |
GenBank mRNA | AK344292.1 | - | 1 - 2 | 134 | 72 | |
AK346059.1 | - | 1 - 2 | 144 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |