Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243447.1 | NP_001230376.1 | 4 - 5 | 462 | 81 | |
GenBank mRNA | CV872394.1 | - | 4 - 5 | 581 | 81 | |
DB786259.1 | - | 4 - 5 | 436 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |