Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNB1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243455.1 | NP_001230384.1 | 4 - 5 | 724 | 100 | |
GenBank mRNA | AK352109.1 | - | 4 - 5 | 575 | 100 | |
CJ029038.1 | - | 4 - 5 | 579 | 100 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Sus scrofa Probe spans exons |