Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243468.1 | NP_001230397.1 | 7 - 8 | 1799 | 72 | |
XM_005663237.2 | XP_005663294.1 | 1473 | 72 | |||
GenBank mRNA | AK345954.1 | - | 7 - 8 | 1799 | 72 | |
AK345986.1 | - | 7 - 8 | 1800 | 72 | ||
DY429032.1 | - | 1 - 2 | 99 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |