Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC46A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243471.1 | NP_001230400.1 | 1 - 2 | 359 | 88 | |
XM_021065096.1 | XP_020920755.1 | 385 | 88 | |||
XM_021065098.1 | XP_020920757.1 | 388 | 88 | |||
GenBank mRNA | AK233023.1 | - | 1 - 2 | 358 | 88 | |
AK345242.1 | - | 1 - 2 | 358 | 88 | ||
BP137848.1 | - | 1 - 2 | 359 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |