Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243478.1 | NP_001230407.1 | 2 - 3 | 362 | 63 | |
GenBank mRNA | AK231488.1 | - | 2 - 3 | 364 | 63 | |
AK240399.1 | - | 2 - 3 | 281 | 63 | ||
AK346690.1 | - | 2 - 3 | 281 | 63 | ||
AY610201.1 | - | 2 - 3 | 338 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |