Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243490.1 | NP_001230419.1 | 3 - 4 | 1153 | 62 | |
GenBank mRNA | AK237777.1 | - | 3 - 4 | 1095 | 62 | |
AK395851.1 | - | 5 - 6 | 1248 | 62 | ||
AK400323.1 | - | 4 - 5 | 1202 | 62 | ||
EW623074.2 | - | 3 - 4 | 386 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |