Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243490.1 | NP_001230419.1 | 4 - 5 | 1202 | 83 | |
GenBank mRNA | AK237777.1 | - | 4 - 5 | 1144 | 83 | |
AK395851.1 | - | 6 - 7 | 1297 | 83 | ||
AK400323.1 | - | 5 - 6 | 1251 | 83 | ||
EW623074.2 | - | 4 - 5 | 435 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |