Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243516.1 | NP_001230445.1 | 2 - 3 | 325 | 91 | |
XM_005671062.3 | XP_005671119.1 | 667 | 91 | |||
XM_013983315.2 | XP_013838769.1 | 301 | 91 | |||
GenBank mRNA | AK344419.1 | - | 2 - 3 | 325 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |