Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243545.1 | NP_001230474.1 | 2 - 3 | 326 | 90 | |
XM_005665556.3 | XP_005665613.2 | 358 | 90 | |||
XM_021098043.1 | XP_020953702.1 | 358 | 90 | |||
GenBank mRNA | AK347131.1 | - | 2 - 3 | 305 | 90 | |
BG833879.1 | - | 1 - 2 | 232 | 90 | ||
FD597021.1 | - | 2 - 3 | 326 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |