Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YDJC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243562.1 | NP_001230491.1 | 4 - 5 | 615 | 56 | |
GenBank mRNA | AK349979.1 | - | 4 - 5 | 615 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons |