Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPPED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243581.1 | NP_001230510.1 | 1 - 2 | 97 | 73 | |
GenBank mRNA | AK232383.1 | - | 1 - 2 | 125 | 73 | |
AK232873.1 | - | 1 - 2 | 106 | 73 | ||
AK345202.1 | - | 1 - 2 | 97 | 73 | ||
AK394535.1 | - | 1 - 2 | 158 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |