Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243604.1 | NP_001230533.1 | 4 - 5 | 512 | 99 | |
XM_005656927.3 | XP_005656984.1 | 520 | 99 | |||
XM_005656929.3 | XP_005656986.1 | 485 | 99 | |||
XM_005656930.3 | XP_005656987.1 | 484 | 99 | |||
XM_005656931.3 | XP_005656988.1 | 444 | 99 | |||
XM_013989897.2 | XP_013845351.1 | 446 | 99 | |||
XM_021066162.1 | XP_020921821.1 | 661 | 99 | |||
GenBank mRNA | AK238125.1 | - | 3 - 4 | 759 | 99 | |
AY609674.1 | - | 4 - 5 | 512 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |