Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243604.1 | NP_001230533.1 | 5 - 6 | 685 | 107 | |
XM_005656927.3 | XP_005656984.1 | 693 | 107 | |||
XM_005656928.3 | XP_005656985.1 | 666 | 107 | |||
XM_005656929.3 | XP_005656986.1 | 658 | 107 | |||
XM_005656930.3 | XP_005656987.1 | 657 | 107 | |||
XM_005656931.3 | XP_005656988.1 | 617 | 107 | |||
XM_013989897.2 | XP_013845351.1 | 619 | 107 | |||
XM_021066162.1 | XP_020921821.1 | 834 | 107 | |||
XM_021066163.1 | XP_020921822.1 | 404 | 107 | |||
XM_021066164.1 | XP_020921823.1 | 311 | 107 | |||
GenBank mRNA | AK238125.1 | - | 4 - 5 | 932 | 107 | |
AY609674.1 | - | 5 - 6 | 685 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |