Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243622.1 | NP_001230551.1 | 3 - 4 | 481 | 109 | |
XM_021100397.1 | XP_020956056.1 | 629 | 109 | |||
XM_021100398.1 | XP_020956057.1 | 637 | 109 | |||
GenBank mRNA | AK232809.1 | - | 3 - 4 | 481 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |