Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243649.1 | NP_001230578.1 | 1 - 2 | 236 | 91 | |
XM_013996774.2 | XP_013852228.1 | 1874 | 91 | |||
GenBank mRNA | AK347101.1 | - | 1 - 2 | 236 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |