Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRA10AC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243654.1 | NP_001230583.1 | 7 - 8 | 605 | 108 | |
XM_005671290.3 | XP_005671347.1 | 669 | 108 | |||
XM_005671293.3 | XP_005671350.1 | 697 | 108 | |||
XM_005671294.3 | XP_005671351.1 | 634 | 108 | |||
XM_013983566.2 | XP_013839020.1 | 521 | 108 | |||
XM_013983569.2 | XP_013839023.1 | 507 | 108 | |||
XM_021072007.1 | XP_020927666.1 | 669 | 108 | |||
GenBank mRNA | AK235945.1 | - | 7 - 8 | 605 | 108 | |
AK346001.1 | - | 8 - 9 | 633 | 108 | ||
AK347603.1 | - | 7 - 8 | 570 | 108 | ||
AK390487.1 | - | 7 - 8 | 617 | 108 | ||
AY610209.1 | - | 7 - 8 | 559 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |