Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM216B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243662.1 | NP_001230591.1 | 3 - 4 | 273 | 79 | |
XM_005656846.3 | XP_005656903.1 | 230 | 79 | |||
XM_005656847.3 | XP_005656904.1 | 230 | 79 | |||
XM_013989774.2 | XP_013845228.1 | 359 | 79 | |||
XM_021065104.1 | XP_020920763.1 | 278 | 79 | |||
XM_021065105.1 | XP_020920764.1 | 419 | 79 | |||
XM_021065106.1 | XP_020920765.1 | 413 | 79 | |||
XM_021065107.1 | XP_020920766.1 | 431 | 79 | |||
XM_021065109.1 | XP_020920768.1 | 259 | 79 | |||
GenBank mRNA | GT640835.1 | - | 2 - 3 | 213 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |