Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243668.1 | NP_001230597.1 | 7 - 8 | 822 | 60 | |
XM_005663288.3 | XP_005663345.1 | 1265 | 60 | |||
XM_005663289.3 | XP_005663346.1 | 962 | 60 | |||
XM_005663290.3 | XP_005663347.1 | 842 | 60 | |||
XM_013996967.2 | XP_013852421.1 | 1444 | 60 | |||
XM_013996968.2 | XP_013852422.1 | 1439 | 60 | |||
XM_013996969.2 | XP_013852423.1 | 1376 | 60 | |||
XM_013996970.2 | XP_013852424.1 | 1352 | 60 | |||
XM_013996971.2 | XP_013852425.1 | 1295 | 60 | |||
XM_013996974.2 | XP_013852428.1 | 916 | 60 | |||
XM_021088598.1 | XP_020944257.1 | 1423 | 60 | |||
XM_021088599.1 | XP_020944258.1 | 1127 | 60 | |||
XM_021088600.1 | XP_020944259.1 | 1274 | 60 | |||
XM_021088601.1 | XP_020944260.1 | 941 | 60 | |||
GenBank mRNA | AK349829.1 | - | 6 - 7 | 753 | 60 | |
AK389884.1 | - | 7 - 8 | 880 | 60 | ||
AK399910.1 | - | 2 - 3 | 248 | 60 | ||
EU649704.1 | - | 6 - 7 | 771 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |