Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243675.1 | NP_001230604.1 | 8 - 9 | 823 | 82 | |
GenBank mRNA | AJ679671.1 | - | 2 - 3 | 145 | 82 | |
AK230481.1 | - | 8 - 9 | 737 | 82 | ||
AK390217.1 | - | 8 - 9 | 821 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |