Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243691.1 | NP_001230620.1 | 6 - 7 | 1188 | 70 | |
GenBank mRNA | AK349998.1 | - | 6 - 7 | 1188 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |