Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243703.1 | NP_001230632.1 | 2 - 3 | 377 | 67 | |
XM_005673417.3 | XP_005673474.1 | 421 | 67 | |||
XM_005673418.3 | XP_005673475.1 | 421 | 67 | |||
GenBank mRNA | AK351224.1 | - | 2 - 3 | 377 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |