Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243709.1 | NP_001230638.1 | 5 - 6 | 879 | 59 | |
GenBank mRNA | AK351364.1 | - | 5 - 6 | 879 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |