Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243713.1 | NP_001230642.1 | 1 - 2 | 129 | 139 | |
GenBank mRNA | AK347703.1 | - | 1 - 2 | 129 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |